missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylphosphinic chloride, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.
Solubility
Decomposes with water.
Notes
Moisture Sensitive. Store away from strong bases, moisture. Incompatible with oxidizing agents,, Amines, Alcohols.
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.
Solubility
Decomposes with water.
Notes
Moisture Sensitive. Store away from strong bases, moisture. Incompatible with oxidizing agents,, Amines, Alcohols.
Specifications
Melting Point | ∼20°C |
Density | 1.242 |
Boiling Point | 210°C to 212°C (8 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.609 |
Quantity | 5 g |
UN Number | UN3265 |
Sensitivity | Moisture sensitive |
Solubility Information | Decomposes with water. |
Formula Weight | 236.64 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'