missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinic acid, 99%

Product Code. 11464613
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464613 100 g 100g
11444613 5 g 5g
11454613 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464613 Supplier Thermo Scientific Alfa Aesar Supplier No. A11563.22
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1707-03-5 | C12H11O2P | 218.19 g/mol

Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1707-03-5
Molecular Formula C12H11O2P
Molecular Weight (g/mol) 218.19
MDL Number MFCD00002132
InChI Key BEQVQKJCLJBTKZ-UHFFFAOYSA-N
Synonym phosphinic acid, diphenyl, hydroxydiphenylphosphine oxide, diphenylphosphinicacid, diphenyl phosphinic acid, diphenyl-phosphinic acid, acmc-1boli, di phenyl phosphinic acid, p,p-diphenylphosphinic acid, ksc180o2h, phosphinicacid, p,p-diphenyl
PubChem CID 15567
ChEBI CHEBI:37832
IUPAC Name diphenylphosphinic acid
SMILES OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 193°C to 197°C
Quantity 100 g
Beilstein 2804567
Solubility Information Soluble in water.
Formula Weight 218.19
Percent Purity 99%
Chemical Name or Material Diphenylphosphinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.