missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphine oxide, 97%

Product Code. 11484557
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484557 1 g 1g
11494557 5 g 5g
11404567 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484557 Supplier Thermo Scientific Alfa Aesar Supplier No. B21101.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4559-70-0 | C12H11OP | 202.19 g/mol

Used in the preparation of triarylphosphine oxides, and as asymmetric catalysts, preparation of alkenyldiphenylphosphine oxides, and Horner-Wittig reagents. It is a ligand for Buchwald-Hartwig Cross Coupling Reaction, Hydrophosphonylations and Suzuki-Miyaura Coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of triarylphosphine oxides, and as asymmetric catalysts, preparation of alkenyldiphenylphosphine oxides, and Horner-Wittig reagents. It is a ligand for Buchwald-Hartwig Cross Coupling Reaction, Hydrophosphonylations and Suzuki-Miyaura Coupling.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4559-70-0
Molecular Formula C12H11OP
Molecular Weight (g/mol) 202.19
MDL Number MFCD00002079
InChI Key ASUOLLHGALPRFK-UHFFFAOYSA-N
Synonym diphenylphosphine oxide, phosphine oxide, diphenyl, diphenyl phosphine oxide, hpoph2, asuollhgalprfk-uhfffaoysa-n, dppo, diphenylphosphane oxide, phenylphosphonoylbenzene, diphenylphosphino-1-one, pubchem23810
PubChem CID 6327869
IUPAC Name oxo(diphenyl)phosphanium
SMILES O=P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 51°C to 56°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 202.2
Percent Purity 97%
Chemical Name or Material Diphenylphosphine oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.