missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinamide, 98%

Product Code. 11417997
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417997 5 g 5g
11427997 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417997 Supplier Thermo Scientific Alfa Aesar Supplier No. A16657.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5994-87-6 | C12H12NOP | 217.21 g/mol

Diphenylphosphinamide is used to furnish 5H-oxazol-4-ones with diastereoselectivity and enantioselectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinamide is used to furnish 5H-oxazol-4-ones with diastereoselectivity and enantioselectivity.

Solubility
Soluble in chloroform, nitromethane, nitrobenzene.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5994-87-6
Molecular Formula C12H12NOP
Molecular Weight (g/mol) 217.21
MDL Number MFCD00014766
InChI Key RIGIWEGXTTUCIQ-UHFFFAOYSA-N
Synonym diphenylphosphinamide, p,p-diphenylphosphinic amide, amino phenyl phosphoryl benzene, amino phenyl phosphoroso benzene, diphenyl phosphinic acid amide, azanyl phenyl phosphoryl benzene
PubChem CID 853560
SMILES NP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 160°C to 162°C
Quantity 5 g
Solubility Information Soluble in chloroform,nitromethane,nitrobenzene.
IUPAC Name [amino(phenyl)phosphoroso]benzene
Formula Weight 217.21
Percent Purity 98%
Chemical Name or Material Diphenylphosphinamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.