missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethanone Hydrazone, TRC

Product Code. 30373822
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373822 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373822 Supplier Toronto Research Chemicals Supplier No. TRCD491685100G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzophenone hydrazone
CAS 5350-57-2
Molecular Formula C13 H12 N2
Quantity 100 g
Synonym Benzophenone Hydrazone, Diphenyl Ketone Hydrazone, Diphenyl methanone Hydrazone, N-(Diphenylmethylene)hydrazine, NSC 43
InChI Formula InChI=1S/C13H12N2/c14-15-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,14H2
SMILES NN=C(c1ccccc1)c2ccccc2
IUPAC Name benzhydrylidenehydrazine
Molecular Weight (g/mol) 196.25
Formula Weight 196.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.