missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylmethanone (Benzophenone), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Diphenylmethanone (Benzophenone)
Specifications
Specifications
| Chemical Name or Material | Benzophenone |
| CAS | 119-61-9 |
| Analyte or Component Names | Benzophenone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H10 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzophenone, Diphenylmethanone, Hydrocodone Hydrogen Tartrate 2.5-Hydrate Imp. H (EP), Phenytoin Imp. A (EP), Phenytoin Sodium Imp. A (EP), Dimenhydrinate Imp. J (EP), Diphenhydramine Hydrochloride Imp. E (EP), Diphenhydramine Imp. E (EP), Hydrocodone Imp. H (EP), SKF-3353, Ph Eur Diphenhydramine HCl Impurity E, Dimenhydrinate Impurity J, Diphenhydramine Hydrochloride Impurity E, Hydrocodone Hydrogen Tartrate 2.5-Hydrate Impurity H, Phenytoin Impurity A, Phenytoin Sodium Impurity A, Diphenhydramine Impurity E, Hydrocodone Impurity H, Phenytoin Impurity A |
| InChI Formula | InChI=1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H |
| SMILES | O=C(c1ccccc1)c2ccccc2 |
| IUPAC Name | diphenylmethanone |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction