missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethanol (Benzhydrol), Mikromol™

Product Code. 17971316
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971316 25 mg 25mg
17954286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971316 Supplier Mikromol Supplier No. MM0490.020025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diphenylmethanol (Benzhydrol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylmethanol
CAS 91-01-0
Analyte or Component Names Diphenylmethanol
Shipping Condition Room Temperature
Molecular Formula C13 H12 O
Certifications/Compliance ISO 17025
Synonym Cyclizine Hydrochloride Imp. B (EP), Cyclizine Imp. B (EP), Dimenhydrinate Imp. I (EP), Diphenhydramine Hydrochloride Imp. D (EP), Diphenhydramine Imp. D (EP), Ebastine Imp. A (EP), Diphenylmethanol, Benzhydrol, Cyclizine Hydrochloride Impurity B, Dimenhydrinate Impurity I, Diphenhydramine Hydrochloride Impurity D, Ebastine Impurity A, Cyclizine Impurity B, Diphenhydramine Impurity D
InChI Formula InChI=1S/C13H12O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13-14H
SMILES OC(c1ccccc1)c2ccccc2
IUPAC Name diphenylmethanol
Molecular Weight (g/mol) 184.23
Quantity 25 mg
Formula Weight 184.0888 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.