missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethane, 99+%

Product Code. 11444787
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444787 100 g 100g
11454787 500 g 500g
11464787 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444787 Supplier Thermo Scientific Alfa Aesar Supplier No. B21256.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 101-81-5 | C13H12 | 168.24 g/mol

Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

Solubility
Soluble in ethanol, ether, benzene and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-81-5
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00004781
InChI Key CZZYITDELCSZES-UHFFFAOYSA-N
Synonym diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
PubChem CID 7580
ChEBI CHEBI:38884
IUPAC Name benzylbenzene
SMILES C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 22°C to 26°C
Density 1.01
Boiling Point 262°C to 264°C
Flash Point 130°C (266°F)
Refractive Index 1.577
Quantity 100 g
UN Number UN3077
Beilstein 1904982
Merck Index 14,3328
Solubility Information Soluble in ethanol,ether,benzene and chloroform.
Formula Weight 168.24
Percent Purity ≥99%
Chemical Name or Material Diphenylmethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.