missing translation for 'onlineSavingsMsg'
Learn More

Diphenyliodonium hexafluorophosphate, 98%

Product Code. 11409066
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11409066

Brand: Thermo Scientific Alfa Aesar A17934.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58109-40-3 | C12H10I | 281.12 g/mol

Diphenyliodonium hexafluorophosphate is used as cationic photoinitiator and photoacid generator. It acts as a catalyst for the photochemical polymerization of various monomers. Further, it used as arylating reagent as well as an oxidant in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyliodonium hexafluorophosphate is used as cationic photoinitiator and photoacid generator. It acts as a catalyst for the photochemical polymerization of various monomers. Further, it used as arylating reagent as well as an oxidant in organic synthesis.

Solubility
Soluble in tetrahydrofuran, ethyl acetate, methanol, acetonitrile, dimethylformamide, dimethyl carbonate. Sparingly soluble in chloroform and dichloromethane. Insoluble in pentanes, hexanes, toluene and diethyl ether.

Notes
Light sensitive and hygroscopic. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58109-40-3
Molecular Formula C12H10I
Molecular Weight (g/mol) 281.12
MDL Number MFCD00061398
InChI Key OZLBDYMWFAHSOQ-UHFFFAOYSA-N
Synonym diphenyliodonium hexafluorophosphate, diphenyliodanium hexafluorophosphate, diphenyliodonium hexafluorophosphate v, iodonium, diphenyl-, hexafluorophosphate 1-, iodonium, diphenyl-, hexafluorophosphate 1-1:1, diphenyliodonium pf6, dpihfp, acmc-209m2o, phi + pf6-, diphenyliodanium; hexafluoro-$l^ 5-phosphanuide
PubChem CID 2737136
IUPAC Name diphenyliodanium
SMILES [I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 139°C to 144°C
Quantity 250 g
UN Number UN2811
Sensitivity Light sensitive
Solubility Information Soluble in tetrahydrofuran,ethyl acetate,methanol,acetonitrile,dimethylformamide,dimethyl carbonate. Sparingly soluble in chloroform and dichloromethane. Insoluble in pentanes,hexanes,toluene and diethyl ether.
Formula Weight 426.08
Percent Purity 98%
Chemical Name or Material Diphenyliodonium hexafluorophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.