missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride, MedChemExpress

Product Code. 16439776
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16439776

Brand: Medchem Express HY100965

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Diphenyleneiodonium chloride is a NADPH oxidase (NOX) inhibitor and also functions as a TRPA1 activator with an EC50 of 1 to 3 μM. Diphenyleneiodonium chloride selectively inhibits intracellular reactive oxygen species.

Diphenyleneiodonium chloride is a NADPH oxidase (NOX) inhibitor and also functions as a TRPA1 activator with an EC50 of 1 to 3 μM. Diphenyleneiodonium chloride selectively inhibits intracellular reactive oxygen species.

Specifications

Chemical Name or Material Diphenyleneiodonium chloride
Physical Form Solid
CAS 4673-26-1
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C12H8ClI
For Use With (Application) COVID-19-immunoregulation
Synonym DPI
Solubility Information DMSO : 6 mg/mL (19.07 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES C12=CC=CC=C1C3=CC=CC=C3[I+]2.[Cl-]
Molecular Weight (g/mol) 314.55
Formula Weight 314.55
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.