missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride, MedChemExpress

Product Code. 16443228
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443228 10 mg 10mg
16439776 50 mg 50mg
16424496 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16443228 Supplier Medchem Express Supplier No. HY100965
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diphenyleneiodonium chloride is a NADPH oxidase (NOX) inhibitor and also functions as a TRPA1 activator with an EC50 of 1 to 3 μM. Diphenyleneiodonium chloride selectively inhibits intracellular reactive oxygen species.

Diphenyleneiodonium chloride is a NADPH oxidase (NOX) inhibitor and also functions as a TRPA1 activator with an EC50 of 1 to 3 μM. Diphenyleneiodonium chloride selectively inhibits intracellular reactive oxygen species.

Specifications

Chemical Name or Material Diphenyleneiodonium chloride
Physical Form Solid
CAS 4673-26-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C12H8ClI
For Use With (Application) COVID-19-immunoregulation
Synonym DPI
Solubility Information DMSO : 6 mg/mL (19.07 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES C12=CC=CC=C1C3=CC=CC=C3[I+]2.[Cl-]
Molecular Weight (g/mol) 314.55
Formula Weight 314.55
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.