missing translation for 'onlineSavingsMsg'
Learn More

Diphenyldithiophosphonic acid

Product Code. 11333397
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333397 10 g 10g
11323397 0.5 g 0.5g
11343397 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11333397 Supplier Thermo Scientific Alfa Aesar Supplier No. 017758.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1015-38-9 | C12H11PS2 | 250.31 g/mol

Diphenyldithiophosphonic acid is used in place of phenyl phosphate for acylation of nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1015-38-9
Molecular Formula C12H11PS2
Molecular Weight (g/mol) 250.31
MDL Number MFCD00014457
InChI Key CLUOCCWZZAGLPM-UHFFFAOYSA-N
Synonym diphenylphosphinodithioic acid, diphenyldithiophosphonic acid, diphenyldithiophosphinic acid, phosphinodithioic acid, diphenyl, diphenyl sulfanyl-??-phosphanethione, dithiodiphenylphosphinic acid, diphenyl-sulfanyl-sulfanylidene, diphenyl sulfanyl-lambda-phosphanethione, diphenyl-sulfanyl-sulfanylidenephosphorane, diphenyl sulfanyl-$l; 5-phosphanethione
PubChem CID 129564
IUPAC Name diphenyl-sulfanyl-sulfanylidene-$l^{5}-phosphane
SMILES SP(=S)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 55°C to 56°C
Quantity 10 g
Formula Weight 250.31
Physical Form Crystalline
Chemical Name or Material Diphenyldithiophosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.