missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyldichlorosilane, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Diphenyldichlorosilane |
| CAS | 80-10-4 |
| Molecular Formula | C12H10Cl2Si |
| Quantity | 5 g |
| Synonym | Dichlorodiphenylsilane, Diphenylsilicon Dichloride, Diphenylsilyl Dichloride, NSC 77110, TSL 8062 |
| InChI Formula | InChI=1S/C12H10Cl2Si/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H |
| SMILES | Cl[Si](Cl)(c1ccccc1)c2ccccc2 |
| IUPAC Name | dichloro(diphenyl)silane |
| Molecular Weight (g/mol) | 253.2 |
| Formula Weight | 251.9929 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction