missing translation for 'onlineSavingsMsg'
Learn More

Diphenyldichlorosilane, 97%, Thermo Scientific Chemicals

Product Code. 11419932
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store away from water/moisture, oxidizing agents and bases. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-10-4
Molecular Formula C12H10Cl2Si
Molecular Weight (g/mol) 253.20
MDL Number MFCD00000489
InChI Key OSXYHAQZDCICNX-UHFFFAOYSA-N
Synonym diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
PubChem CID 6627
IUPAC Name dichloro(diphenyl)silane
SMILES Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 303°C to 304°C
Melting Point -22°C
Quantity 100 g
UN Number UN1769
Beilstein 609882
Flash Point 157°C (314°F)
Solubility Information Decomposes in water.
Formula Weight 253.2
Percent Purity 97%
Odor Pungent
Refractive Index 1.579
Sensitivity Moisture sensitive
Density 1.208
Chemical Name or Material Diphenyldichlorosilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'