missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcyclopropenone, LoGiCal

Product Code. 30460904
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460904 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460904 Supplier LoGiCal Supplier No. LGCFOR3487.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylcyclopropenone
CAS 886-38-4
Analyte or Component Names Diphenylcyclopropenone
Shipping Condition Room Temperature
Molecular Formula C15 H10 O
Certifications/Compliance ISO 9001
Synonym Diphenylcyclopropenone, 2,3-Diphenyl-2-cyclopropen-1-one, Cyclopropenone, diphenyl- (6CI,7CI,8CI), 1,2-Diphenylcyclopropenone, 2,3-Diphenylcyclopropenone, DPC, Diphencyclopropenone, Diphencyprone, NSC 57541
InChI Formula InChI=1S/C15H10O/c16-15-13(11-7-3-1-4-8-11)14(15)12-9-5-2-6-10-12/h1-10H
Solution Type Neat
SMILES O=C1C(=C1c2ccccc2)c3ccccc3
IUPAC Name 2,3-diphenylcycloprop-2-en-1-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 206.24
Quantity 10 mg
Formula Weight 206.0732 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.