missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcyclopropenone, ≥98%, MP Biomedicals™

Product Code. 11431571 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431571 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11431571 Supplier MP Biomedicals Supplier No. 0215952301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 886-38-4
Color Tan
Assay Percent Range ≥98%
Molecular Formula C15H10O
MDL Number MFCD00001311
Quantity 1 g
Synonym diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec
InChI Key HCIBTBXNLVOFER-UHFFFAOYSA-N
SMILES O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name diphenylcycloprop-2-en-1-one
Molecular Weight (g/mol) 206.24
PubChem CID 65057
ChEBI CHEBI:53074
Formula Weight 206.24
Physical Form Powder
Chemical Name or Material Diphenylcyclopropenone
Show More Show Less
compliance-icons
Product Identifier
  • 1GR DIPHENYLCYCLOPROPENONE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Diphenylcyclopropenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.