missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcyclopropenone, 98%

Product Code. 10256230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10256230 1 g Glass bottle 1g
10317740 5 g Glass bottle 5g
11954851 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10256230 Supplier Thermo Scientific Acros Supplier No. 175820010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 886-38-4 | C15H10O | 206.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-38-4
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001311
InChI Key HCIBTBXNLVOFER-UHFFFAOYSA-N
Synonym diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec
PubChem CID 65057
ChEBI CHEBI:53074
IUPAC Name 2,3-diphenylcycloprop-2-en-1-one
SMILES O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 117.0°C to 121.0°C
Color Beige to Orange-Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 07,IV,1736
Merck Index 15,3338
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, ethanol, acetone, chloroform,, dichloromethane, cyclohexane, benzene, hexane,, petroleum ether
Formula Weight 206.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Diphenylcyclopropenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.