missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcarbamyl chloride, 98%

Product Code. 10687522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10687522 25 g Glass Bottle 25g
10256170 100 g Glass Bottle 100g
10494765 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10687522 Supplier Thermo Scientific Acros Supplier No. 167140250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-01-2 | C13H10ClNO | 231.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-01-2
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.68
MDL Number MFCD00000633
InChI Key XNBKKRFABABBPM-UHFFFAOYSA-N
Synonym diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl
PubChem CID 65741
IUPAC Name N,N-diphenylcarbamoyl chloride
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl

Specifications

Melting Point 81.0°C to 85.0°C
Color Beige to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula (C6H5)2NCOCl
Merck Index 15, 3351
Solubility Information Solubility in water: reacts.
Quantity 25 g
Formula Weight 231.68
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Diphenylcarbamyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Diphenylcarbamyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.