missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, TRC

Product Code. 30373303
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30373303 50 g 50g
30381189 5 g -
2 options
This item is not returnable. View return policy

Product Code. 30373303

Brand: Toronto Research Chemicals TRCD47894050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-39-4
Formula Weight 169.0891
InChI Formula InChI=1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H
IUPAC Name N-phenylaniline
Molecular Formula C12 H11 N
Molecular Weight (g/mol) 169.22
SMILES N(c1ccccc1)c2ccccc2
Synonym Benzenamine, N-phenyl-, Diphenylamine (8CI), N-Phenylbenzenamine, Anilinobenzene, Antioxidant DPA, Benzene, (phenylamino)-, DBA, DFA, DPA, N,N-Diphenylamine, N-Phenylaniline, NSC 215210, Naugalube 428L, No-Scald
Chemical Name or Material Diphenylamine

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.