missing translation for 'onlineSavingsMsg'
Learn More

DipHenylamine, Certified AR for Analysis, Fisher Chemical™

Product Code. 10755711 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10755711 250 g 250g
10293180 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10755711 Supplier Fisher Chemical Supplier No. D/4700/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number 3014
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Melting Point 52°C
Boiling Point 302°C
Packaging Glass Jar
Quantity 250 g
Formula Weight 169.23g/mol
Physical Form Solid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diphenylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.