missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetylene, TRC

Product Code. 30382624
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
missing translation for 'unitSize'
10g
Product Code. Quantity unitSize
30382624 10 g 10g
1 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 30382624

missing translation for 'mfr': Toronto Research Chemicals TRCD49339810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylacetylene
CAS 501-65-5
Molecular Formula C14H10
Quantity 10 g
Synonym 1,2-Diphenylacetylene, 1,2-Diphenylethyne, Diphenylacetylene, Diphenylethyne, Diphenylethyne, NSC 5185, Tolan, Tolane
InChI Formula InChI=1S/C14H10/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-10H
SMILES c1ccc(cc1)C#Cc2ccccc2
Molecular Weight (g/mol) 178.23
Formula Weight 178.0783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.