missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylacetyl Chloride, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Diphenylacetyl Chloride |
| CAS | 1871-76-7 |
| Molecular Formula | C14H11ClO |
| Quantity | 5 g |
| Synonym | Diphenylacetyl Acetyl Chloride, Diphenylacetic acid chloride, Diphenylacetyl ChlorideNSC 120906, alpha,alpha-Diphenylacetyl Chloride, alpha-Phenylbenzeneacetyl Chloride |
| InChI Formula | InChI=1S/C14H11ClO/c15-14(16)13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H |
| SMILES | ClC(=O)C(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2,2-diphenylacetyl chloride |
| Molecular Weight (g/mol) | 230.69 |
| Formula Weight | 230.0498 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction