missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetyl chloride, 97.5%

Product Code. 10375221 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10375221

Brand: Thermo Scientific Acros 292020050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1871-76-7 | C14H11ClO | 230.691 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-76-7
MDL Number MFCD00013655
InChI Key MSYLETHDEIJMAF-UHFFFAOYSA-N
Synonym diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride
PubChem CID 74637
IUPAC Name 2,2-diphenylacetyl chloride
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl

Specifications

Melting Point 51°C to 56°C
Color Yellow
Boiling Point 175°C to 176°C (17.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5%
Packaging Glass bottle
Molecular Formula C14H11ClO
Linear Formula (C6H5)2CHCOCl
Quantity 5 g
Beilstein 09, 674
Merck Index 15, 3346
Solubility Information Solubility in water: may decompose
Molecular Weight (g/mol) 230.69
Formula Weight 230.69
Percent Purity 97.50%
Physical Form Crystalline Powder
Chemical Name or Material Diphenylacetyl chloride, 97.5%
Show More Show Less
compliance-icons
Product Identifier
  • Diphenylacetylchloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Diphenylacetylchloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.