missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetyl chloride, 90+%

Product Code. 11342685
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11342685

Brand: Thermo Scientific Alfa Aesar L00171.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1871-76-7 | C14H11ClO | 230.691 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-76-7
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.691
MDL Number MFCD00013655
InChI Key MSYLETHDEIJMAF-UHFFFAOYSA-N
Synonym diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride
PubChem CID 74637
IUPAC Name 2,2-diphenylacetyl chloride
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl

Specifications

Melting Point 50°C to 56°C
Boiling Point 175°C to 176°C (17 mmHg)
Quantity 10 g
UN Number UN3261
Beilstein 1211588
Sensitivity Moisture sensitive
Merck Index 14,3316
Formula Weight 230.69
Percent Purity ≥90%
Chemical Name or Material Diphenylacetyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.