missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetonitrile, TRC

Product Code. 30010892
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010892 2.5 g 2.5g
30136222 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010892 Supplier Toronto Research Chemicals Supplier No. TRCD4894502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-29-3
Formula Weight 193.0891
InChI Formula InChI=1S/C14H11N/c15-11-14(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,14H
IUPAC Name 2,2-diphenylacetonitrile
Molecular Formula C14 H11 N
Molecular Weight (g/mol) 193.24
SMILES N#CC(c1ccccc1)c2ccccc2
Synonym Diphenylacetonitrile, Methadone Hydrochoride Imp. E (EP)
Chemical Name or Material Diphenylacetonitrile

Specifications

Quantity 2.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.