missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetonitrile, Mikromol™

Product Code. 30050929
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050929 100 mg 100mg
30051819 25 mg 25mg
30460533 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30050929 Supplier Mikromol Supplier No. MM0531.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diphenylacetonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylacetonitrile
CAS 86-29-3
Analyte or Component Names Diphenylacetonitrile
Shipping Condition Room Temperature
Molecular Formula C14 H11 N
Certifications/Compliance ISO 17025
Synonym Methadone Hydrochoride Imp. E (EP), Methadone Imp. E (EP), Diphenylacetonitrile, Methadone Hydrochloride Impurity E, Methadone Impurity E
InChI Formula InChI=1S/C14H11N/c15-11-14(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,14H
SMILES N#CC(c1ccccc1)c2ccccc2
IUPAC Name 2,2-diphenylacetonitrile
Molecular Weight (g/mol) 193.24
Quantity 100 mg
Formula Weight 193.0891 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.