missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetic Acid, TRC

Product Code. 30383147
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383147 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383147 Supplier Toronto Research Chemicals Supplier No. TRCD4869855G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylacetic Acid
CAS 117-34-0
Molecular Formula C14H12O2
Quantity 5 g
Synonym Acetic acid, diphenyl- (6CI,7CI,8CI), α-Phenylbenzeneacetic acid, 2,2-Diphenylacetic acid, Diphenylacetic acid, Diphenylethanoic acid, NSC 120417, NSC 8742, α,α-Diphenylacetic acid, Diphenylacetic Acid
InChI Formula InChI=1S/C14H12O2/c15-14(16)13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H,(H,15,16)
SMILES OC(=O)C(c1ccccc1)c2ccccc2
IUPAC Name 2,2-diphenylacetic acid
Molecular Weight (g/mol) 212.24
Formula Weight 212.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.