missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetic acid, 99%

Product Code. 11430393
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430393 100 g 100g
11440393 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430393 Supplier Thermo Scientific Alfa Aesar Supplier No. A11489.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 117-34-0 | C14H12O2 | 212.248 g/mol

Kinetic resolution of racemic 1-heteroarylalkanols by asymmetric esterification using diphenylacetic acid with pivalic anhydride and a chiral acyl-transfer catalyst is described. Molecular crystal of acridine and diphenylacetic acid and its absolute asymmetric photodecarboxylating condensation generates chirality in a two-component. The activation parameters for the racemization of a series of ortho-alkyl-substituted diphenylacetic acids are determined.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Kinetic resolution of racemic 1-heteroarylalkanols by asymmetric esterification using diphenylacetic acid with pivalic anhydride and a chiral acyl-transfer catalyst is described. Molecular crystal of acridine and diphenylacetic acid and its absolute asymmetric photodecarboxylating condensation generates chirality in a two-component. The activation parameters for the racemization of a series of ortho-alkyl-substituted diphenylacetic acids are determined.

Solubility
Slightly soluble in water(0.13g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-34-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00004251
InChI Key PYHXGXCGESYPCW-UHFFFAOYSA-N
Synonym diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl
PubChem CID 8333
ChEBI CHEBI:41967
IUPAC Name 2,2-diphenylacetic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 146°C to 148°C
Boiling Point 195°C (25 mm)
Assay Percent Range 99%
Quantity 100 g
Beilstein 1910978
Merck Index 14,3316
Solubility Information Slightly soluble in water(0.13g/L).
Formula Weight 212.25
Percent Purity 99%
Chemical Name or Material Diphenylacetic Acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.