missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Sulfide, TRC

Product Code. 30379536
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379536 25 g 25g
30375633 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379536 Supplier Toronto Research Chemicals Supplier No. TRCD49198025G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenyl sulfide
CAS 139-66-2
Molecular Formula C12 H10 S
Quantity 25 g
Synonym Phenyl sulfide (8CI), 1,1'-Thiobis[benzene], Diphenyl monosulfide, Diphenyl sulfide, Diphenyl sulphide, Diphenyl thioether, Diphenylmercaptan, Diphenylsulfane, NSC 4568, Phenylthiobenzene
InChI Formula InChI=1S/C12H10S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H
SMILES S(c1ccccc1)c2ccccc2
IUPAC Name phenylsulfanylbenzene
Molecular Weight (g/mol) 186.27
Formula Weight 186.0503
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.