missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phthalate, 98%

Product Code. p-7021916
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457107 25 g 25g
11467107 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457107 Supplier Thermo Scientific Alfa Aesar Supplier No. A14449.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84-62-8 | C20H14O4 | 318.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-62-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00003038
InChI Key DWNAQMUDCDVSLT-UHFFFAOYSA-N
Synonym diphenyl phthalate, phenyl phthalate, phthalic acid, diphenyl ester, diphenylphthalate, 1,2-benzenedicarboxylic acid, diphenyl ester, phthalic acid diphenyl ester, caswell no. 399b, unii-bu20109xhv, 1,2-benzenedicarboxylic acid, 1,2-diphenyl ester, epa pesticide chemical code 017001
PubChem CID 6778
ChEBI CHEBI:60819
IUPAC Name diphenyl benzene-1,2-dicarboxylate
SMILES O=C(OC1=CC=CC=C1)C1=CC=CC=C1C(=O)OC1=CC=CC=C1

Specifications

Melting Point 74°C to 76°C
Density 1.28
Boiling Point 400°C to 405°C
Flash Point 224°C (435°F)
Odor Odorless
Quantity 25 g
Beilstein 2473390
Merck Index 14,7306
Formula Weight 318.33
Percent Purity 98%
Chemical Name or Material Diphenyl phthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.