missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phthalate-3,4,5,6-d4, TRC

Product Code. 30381351
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381351 1 mg -
30377167 10 mg -
30387965 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381351 Supplier Toronto Research Chemicals Supplier No. TRCD4915911MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1398065-61-6
Molecular Formula C202H4H10O4
Molecular Weight (g/mol) 322.35
Synonym 1,2-Benzenedicarboxylic acid-3,4,5,6-d4, 1,2-diphenyl ester, Phthalic acid, bis-phenyl ester D4, Phthalic Acid, Bis-phenyl Ester D4 (phenyl D4)
IUPAC Name diphenyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
SMILES [2H]c1c([2H])c([2H])c(C(=O)Oc2ccccc2)c(C(=O)Oc3ccccc3)c1[2H]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H14O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h1-14H/i7D,8D,13D,14D
Formula Weight 322.1143
Chemical Name or Material Phthalic acid, bis-phenyl ester D4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.