missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phosphite, TRC

Product Code. 30373810
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373810 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373810 Supplier Toronto Research Chemicals Supplier No. TRCD49224550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenyl Phosphite
CAS 4712-55-4
Molecular Formula C12H11O3P
Quantity 50 g
Synonym Diphenoxyphosphine Oxide, Diphenyl Hydrogen Phosphite, Diphenyl PhosphiteDiphenyl Phosphonate, Doverphos 213, JP 260, NSC 43786, Weston DPP
InChI Formula InChI=1S/C12H11O3P/c13-16(14-11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h1-10,16H
SMILES O=P(Oc1ccccc1)Oc2ccccc2
IUPAC Name phenoxyphosphonoyloxybenzene
Molecular Weight (g/mol) 234.19
Formula Weight 234.0446
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.