missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphite, contains varying amounts of phenol and (C6H5O)3P

Product Code. 10448450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10448450 500 g Glass bottle 500g
10386790 5 g Glass bottle 5g
10040790 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10448450 Supplier Thermo Scientific Acros Supplier No. 117395000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4712-55-4 | C12H11O3P | 234.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4712-55-4
Molecular Formula C12H11O3P
Molecular Weight (g/mol) 234.19
MDL Number MFCD00044497
InChI Key CDXVUROVRIFQMV-UHFFFAOYSA-N
Synonym diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
PubChem CID 6327546
IUPAC Name oxo(diphenoxy)phosphanium
SMILES C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2

Specifications

Acidity 15mg KOH/g max.
Melting Point 12°C
Density 1.2170g/mL
Boiling Point 218°C to 219°C (26.0 mmHg)
Flash Point 176°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C6H5O)2P(O)H
Refractive Index 1.5565 to 1.5585
Quantity 500 g
Beilstein 06, I, 94
Fieser 04,210
Specific Gravity 1.217
Solubility Information Solubility in water: insoluble
Formula Weight 234.19
Physical Form Liquid
Chemical Name or Material Diphenyl phosphite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.