missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phosphite, Contains Varying Amounts of Phenol and (C6H5O)3P, Thermo Scientific Chemicals

Catalog Number 10386790
Click to view available options
Quantity:
5 g
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
4712-55-4
C12H11O3P
234.19
MFCD00044497
CDXVUROVRIFQMV-UHFFFAOYSA-N
diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
6327546
oxo(diphenoxy)phosphanium
C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2
This item is not returnable. View return policy
4712-55-4
C12H11O3P
234.19
MFCD00044497
CDXVUROVRIFQMV-UHFFFAOYSA-N
diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
6327546
oxo(diphenoxy)phosphanium
C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 15mg KOH/g max.
Melting Point 12°C
Density 1.2170g/mL
Boiling Point 218°C to 219°C (26.0 mmHg)
Flash Point 176°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C6H5O)2P(O)H
Refractive Index 1.5565 to 1.5585
Quantity 5 g
Beilstein 06, I, 94
Fieser 04,210
Specific Gravity 1.217
Solubility Information Solubility in water: insoluble
Formula Weight 234.19
Physical Form Liquid
Chemical Name or Material Diphenyl phosphite
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'