missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phosphate, TRC

Product Code. 30372820
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372820 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372820 Supplier Toronto Research Chemicals Supplier No. TRCD4920005G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenyl Phosphate
CAS 838-85-7
Purity Grade Notes HPLC
Molecular Formula C12 H11 O4 P
Quantity 5 g
Synonym Diphenyl hydrogen phosphate, Diphenyl phosphate, NSC 6518, Phenyl phosphate ((PhO)2(HO)PO)
InChI Formula InChI=1S/C12H11O4P/c13-17(14,15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H,(H,13,14)
SMILES OP(=O)(Oc1ccccc1)Oc2ccccc2
IUPAC Name diphenyl hydrogen phosphate
Molecular Weight (g/mol) 250.19
Formula Weight 250.0395
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.