missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphate, 97%

Product Code. 11381517
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381517 100 g 100g
11361517 5 g 5g
11371517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381517 Supplier Thermo Scientific Alfa Aesar Supplier No. L02121.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 838-85-7 | C12H11O4P | 250.19 g/mol

Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.

Solubility
Soluble in benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 838-85-7
Molecular Formula C12H11O4P
Molecular Weight (g/mol) 250.19
MDL Number MFCD00003033
InChI Key ASMQGLCHMVWBQR-UHFFFAOYSA-N
Synonym diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
PubChem CID 13282
IUPAC Name diphenyl hydrogen phosphate
SMILES OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Tekniske data

Melting Point 62°C to 70°C
Quantity 100 g
Beilstein 1379164
Solubility Information Soluble in benzene. Insoluble in water.
Formula Weight 250.19
Percent Purity 97%
Chemical Name or Material Diphenyl phosphate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.