missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl methylphenyl phosphate, mixture of isomers, 94%

Product Code. 11359737
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359737 100 mL 100mL
11326049 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359737 Supplier Thermo Scientific Alfa Aesar Supplier No. 042092.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26444-49-5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26444-49-5
MDL Number MFCD00014920
Synonym Phosphoric acid diphenyl methylphenyl ester; Cresyl diphenyl phosphate

Specifications

Melting Point -38°C
Color Yellow
Density 1.21 g/mL
Boiling Point 390°C
Flash Point 232°C (450°F)
Refractive Index 1.563
Quantity 100 mL
UN Number UN3082
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Diphenyl methylphenyl phosphate, Mixture of isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.