missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Methyl Phosphate-d3, TRC

Product Code. 30385020
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385020 10 mg
30384568 100 mg
30381582 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30385020 Supplier Toronto Research Chemicals Supplier No. TRCD49242410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C13 D3 H10 O4 P
Molecular Weight (g/mol) 267.232
Synonym Methoxydiphenoxyphosphine Oxide-d3, Methyl Diphenyl Phosphate-d3
IUPAC Name diphenyl trideuteriomethyl phosphate
SMILES [2H]C([2H])([2H])OP(=O)(Oc1ccccc1)Oc2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H13O4P/c1-15-18(14,16-12-8-4-2-5-9-12)17-13-10-6-3-7-11-13/h2-11H,1H3/i1D3
Formula Weight 267.074
Chemical Name or Material Diphenyl Methyl Phosphate-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.