missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Ditelluride, TRC

Product Code. 30383625
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383625 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383625 Supplier Toronto Research Chemicals Supplier No. TRCD491480100MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenyl Ditelluride
CAS 32294-60-3
Molecular Formula C12H10Te2
Quantity 100 mg
Synonym Diphenyl Ditellurium
InChI Formula InChI=1S/C12H10Te2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H
SMILES [Te]([Te]c1ccccc1)c2ccccc2
IUPAC Name (phenylditellanyl)benzene
Molecular Weight (g/mol) 409.41
Formula Weight 413.8907
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.