missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl ditelluride, 97%

Product Code. 10591141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10591141 1 g Glass bottle 1g
10107853 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10591141 Supplier Thermo Scientific Acros Supplier No. 301840010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32294-60-3 | C12H10Te2 | 409.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32294-60-3
Molecular Formula C12H10Te2
Molecular Weight (g/mol) 409.27
MDL Number MFCD00192106
InChI Key VRLFOXMNTSYGMX-UHFFFAOYSA-N
Synonym diphenyl ditelluride, phenyl ditelluride, ditelluride, diphenyl, diphenylditelluride, ditelluride, diephenyl, phenylditellanyl benzene, diphenylditellane, diphenyl pertelluride, acmc-209hsa, 1,2-diphenylditellane #
PubChem CID 100657
IUPAC Name (phenylditellanyl)benzene
SMILES C1=CC=C(C=C1)[Te][Te]C2=CC=CC=C2

Specifications

Melting Point 64°C to 67°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
UN Number 3284
Solubility Information Solubility in water: insoluble
Formula Weight 409.27
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material Diphenyl ditelluride
Show More Show Less
compliance-icons
Product Identifier
  • Diphenyl ditelluride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.