missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl diselenide, 98%

Product Code. 11431638
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431638 100 g 100g
11411638 5 g 5g
11421638 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431638 Supplier Thermo Scientific Alfa Aesar Supplier No. A13919.22
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1666-13-3 | C12H10Se2 | 312.15 g/mol

Diphenyl diselenide is used in the methoxyselenenylation of alkenes, dihydroxylation of double bonds, hydrothiolation of terminal alkynes. It is used in the synthesis of the unsymmetrical diorganyl selenides, 1-(phenylselenomethyl)vinyl selenides, allylic phenyl selenides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl diselenide is used in the methoxyselenenylation of alkenes, dihydroxylation of double bonds, hydrothiolation of terminal alkynes. It is used in the synthesis of the unsymmetrical diorganyl selenides, 1-(phenylselenomethyl)vinyl selenides, allylic phenyl selenides.

Solubility
Soluble in water, ether.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, halogens, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1666-13-3
Molecular Formula C12H10Se2
Molecular Weight (g/mol) 312.15
MDL Number MFCD00003001
InChI Key YWWZCHLUQSHMCL-UHFFFAOYSA-N
Synonym diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq
PubChem CID 15460
IUPAC Name (phenyldiselanyl)benzene
SMILES [Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 61°C to 63°C
Quantity 100 g
UN Number UN3283
Beilstein 2047179
Solubility Information Soluble in water,ether.
Formula Weight 312.13
Percent Purity 98%
Chemical Name or Material Diphenyl diselenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.