missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Chlorophosphonate, TRC

Product Code. 30378437
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378437 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378437 Supplier Toronto Research Chemicals Supplier No. TRCD48947050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenyl Chlorophosphonate
CAS 2524-64-3
Molecular Formula C12H10ClO3P
Quantity 50 g
Synonym Phenyl Phosphorochloridite, Chlorodiphenoxyphosphine Oxide, Chlorophosphonic Acid Diphenyl Ester, DPC, DPC (flame retardant), DPPC, Diphenoxychlorophosphine Oxide, Diphenyl Chlorophosphate, Diphenyl Phosphochloridate, Diphenyl Phosphorochlorate, Diphenylphosphoric Acid Monochloride, Diphenylphosphoryl Chloride, NSC 43771, O,O-Diphenyl Chlorophosphate, Phenyl Phosphorochloridate ((PhO)2ClPO), Phosphoric Acid Chloride Diphenyl Ester
InChI Formula InChI=1S/C12H10ClO3P/c13-17(14,15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H
SMILES ClP(=O)(Oc1ccccc1)Oc2ccccc2
Molecular Weight (g/mol) 268.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.