missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Chlorophosphonate-d10, TRC

Product Code. 30386755
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386755 100 mg 100mg
30376695 25 mg 25mg
30384917 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386755 Supplier Toronto Research Chemicals Supplier No. TRCD489472100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12 D10 Cl O3 P
Molecular Weight (g/mol) 278.694
Synonym Phenyl Phosphorochloridite-d10, Chlorodiphenoxyphosphine-d10 Oxide, Chlorophosphonic Acid Diphenyl Ester-d10, DPC-d10, DPC-d10 (flame retardant), DPPC-d10, Diphenoxychlorophosphine Oxide, -d10 Diphenyl Chlorophosphate-d10, Diphenyl Phosphochloridate-d10, Diphenyl Phosphorochlorate-d10, Diphenylphosphoric Acid Monochloride-d10, Diphenylphosphoryl Chloride-d10, NSC 43771-d10, O,O-Diphenyl Chlorophosphate-d10, Phenyl Phosphorochloridate ((PhO)2ClPO)-d10, Phosphoric Acid Chloride Diphenyl Ester-d10
IUPAC Name 1-[chloro-(2,3,4,5,6-pentadeuteriophenoxy)phosphoryl]oxy-2,3,4,5,6-pentadeuteriobenzene
SMILES [2H]c1c([2H])c([2H])c(OP(=O)(Cl)Oc2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H10ClO3P/c13-17(14,15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
Formula Weight 278.068
Chemical Name or Material Diphenyl Chlorophosphonate-d10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.