missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl carbonate, 99%

Product Code. 10177653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177653 2.5 kg Plastic bottle 2.5kg
10530751 100 g Plastic bottle 100g
10580581 1 kg Plastic bottle 1kg
11945151 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10177653 Supplier Thermo Scientific Acros Supplier No. 117230025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 102-09-0 | C13H10O3 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-09-0
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
InChI Key ROORDVPLFPIABK-UHFFFAOYSA-N
Synonym carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate
PubChem CID 7597
ChEBI CHEBI:34722
IUPAC Name diphenyl carbonate
SMILES C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2

Specifications

Melting Point 78.0°C to 81.0°C
Color White
Boiling Point 301.0°C to 302.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5O)2CO
Quantity 2.5 kg
Beilstein 06,158
Merck Index 15,3354
Solubility Information Solubility in water: insoluble. Other solubilities: solulbe in acetone, hot alcohol, benzene, carbon, tetrachloride, ether, glacial acetic acid and, other organic solvents
Formula Weight 214.22
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Diphenyl carbonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diphenyl carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.