Learn More
Diphenyl carbonate, 99%, Thermo Scientific Chemicals
£24.20 - £364.00
Chemical Identifiers
CAS | 102-09-0 |
---|---|
Molecular Formula | C13H10O3 |
Molecular Weight (g/mol) | 214.22 |
InChI Key | ROORDVPLFPIABK-UHFFFAOYSA-N |
Synonym | carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate |
PubChem CID | 7597 |
ChEBI | CHEBI:34722 |
IUPAC Name | diphenyl carbonate |
SMILES | C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10530751
|
Thermo Scientific Acros
117231000 |
100 g | Plastic bottle |
£24.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10580581
|
Thermo Scientific Acros
117230010 |
1 kg | Plastic bottle |
£56.40
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10177653
|
Thermo Scientific Acros
117230025 |
2.5 kg | Plastic bottle |
£112.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11945151
|
Thermo Scientific Acros
117230100 |
10 kg | Plastic drum |
£364.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
102-09-0 | |
214.22 | |
carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate | |
CHEBI:34722 | |
C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2 |
C13H10O3 | |
ROORDVPLFPIABK-UHFFFAOYSA-N | |
7597 | |
diphenyl carbonate |
Specifications
102-09-0 | |
White | |
168°C | |
98.5% min. (GC) | |
(C6H5O)2CO | |
15,3354 | |
Solubility in water: insoluble. Other solubilities: solulbe in acetone, hot alcohol, benzene, carbon, tetrachloride, ether, glacial acetic acid and, other organic solvents | |
C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2 | |
214.22 | |
CHEBI:34722 | |
99% | |
Diphenyl carbonate |
78.0°C to 81.0°C | |
301.0°C to 302.0°C | |
Authentic | |
C13H10O3 | |
06,158 | |
carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate | |
ROORDVPLFPIABK-UHFFFAOYSA-N | |
diphenyl carbonate | |
7597 | |
214.22 | |
Crystalline Powder or Flakes |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-005-8
- Diphenyl carbonate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only