Learn More
Diphenyl(4-piperidinyl)methanol, 95%, Thermo Scientific Chemicals
£55.70 - £215.00
Chemical Identifiers
CAS | 115-46-8 |
---|---|
Molecular Formula | C18H21NO |
Molecular Weight (g/mol) | 267.372 |
MDL Number | MFCD00066980 |
InChI Key | ZMISODWVFHHWNR-UHFFFAOYSA-N |
Synonym | azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol |
PubChem CID | 15723 |
IUPAC Name | diphenyl(piperidin-4-yl)methanol |
SMILES | C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
115-46-8 | |
267.372 | |
ZMISODWVFHHWNR-UHFFFAOYSA-N | |
15723 | |
C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O |
C18H21NO | |
MFCD00066980 | |
azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol | |
diphenyl(piperidin-4-yl)methanol |
Specifications
115-46-8 | |
White | |
MFCD00066980 | |
azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol | |
C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O | |
267.372 | |
267.37 | |
Solid |
160°C to 163°C | |
C18H21NO | |
13,895 | |
ZMISODWVFHHWNR-UHFFFAOYSA-N | |
diphenyl(piperidin-4-yl)methanol | |
15723 | |
95% | |
Diphenyl(4-piperidinyl)methanol |
Seguridad y manipulación
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
missing translation for 'einecsNumber' : 204-092-5
missing translation for 'rtecsNumber' : TN0470000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only