missing translation for 'onlineSavingsMsg'
Learn More

α,α-Diphenyl-4-piperidinomethanol, 98+%, Thermo Scientific™

Product Code. 10199921
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 115-46-8
Molecular Formula C18H21NO
Molecular Weight (g/mol) 267.37
MDL Number MFCD00066980
InChI Key ZMISODWVFHHWNR-UHFFFAOYSA-N
Synonym azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol
PubChem CID 15723
IUPAC Name diphenyl(piperidin-4-yl)methanol
SMILES C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 160.0°C to 163.0°C
Color White to Beige
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 13, 895
Formula Weight 267.37
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material α, α-Diphenyl-4-piperidinomethanol, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • a,a-Diphenyl-4-piperidinomethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'