missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl[4-(4-Fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonyl)amino-pyrimidin-5-yl-methyl]phosphine Oxide, TRC

Product Code. 30383583
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383583 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383583 Supplier Toronto Research Chemicals Supplier No. TRCD49160010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenyl[4-(4-Fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonyl)amino-pyrimidin-5-yl-methyl]phosphine Oxide
CAS 289042-10-0
Molecular Formula C28H29FN3O3PS
Quantity 10 g
Synonym Methanesulfonamide, N-[5-[(diphenylphosphinyl)methyl]-4-(4-fluorophenyl)-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-, N-[5-[(Diphenylphosphinyl)methyl]-4-(4-fluorophenyl)-6-(1-methylethyl)-2-pyrimidinyl]-N-methylmethanesulfonamide, Diphenyl[4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonyl)amino-pyrimidin-5-yl-methyl]phosphine Oxide
InChI Formula InChI=1S/C28H29FN3O3PS/c1-20(2)26-25(19-36(33,23-11-7-5-8-12-23)24-13-9-6-10-14-24)27(21-15-17-22(29)18-16-21)31-28(30-26)32(3)37(4,34)35/h5-18,20H,19H2,1-4H3
SMILES CC(C)c1nc(nc(c1CP(=O)(c2ccccc2)c3ccccc3)c4ccc(F)cc4)N(C)S(=O)(=O)C
IUPAC Name N-[5-(diphenylphosphorylmethyl)-4-(4-fluorophenyl)-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
Molecular Weight (g/mol) 537.59
Formula Weight 537.1651
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.