missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-2-pyridylphosphine, 98%

Product Code. 15422458
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422458 25 g 25g
15412458 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422458 Supplier Thermo Scientific Alfa Aesar Supplier No. H56029.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37943-90-1 | C17H14NP | 263.28 g/mol

It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

Solubility
Soluble in toluene. (almost transparency)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37943-90-1
Molecular Formula C17H14NP
Molecular Weight (g/mol) 263.28
MDL Number MFCD00192108
InChI Key SVABQOITNJTVNJ-UHFFFAOYSA-N
Synonym diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
PubChem CID 621893
IUPAC Name diphenyl(pyridin-2-yl)phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3

Specifications

Melting Point 82°C to 84°C
Quantity 25 g
Solubility Information Soluble in toluene. (almost transparency)
Formula Weight 263.27
Percent Purity 98%
Chemical Name or Material Diphenyl-2-pyridylphosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.