missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl(2,4,6-trimethylbenzoyl)phosphine Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide |
| CAS | 75980-60-8 |
| Molecular Formula | C22 H21 O2 P |
| Quantity | 5 g |
| Synonym | Phosphine oxide, diphenyl(2,4,6-trimethylbenzoyl)- (9CI), (Diphenylphosphinyl)(2,4,6-trimethylphenyl)methanone, (2,4,6-Trimethylbenzoyl)diphenylphosphine oxide, Additol TPO, Chemcure TPO, Chivacure TPO, Ciba TPO, D 3358, D-TPO, DCTPO, Darocur TPO, Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide, Doublecure TPO, Esacure TPO, Genocure LTM, Genocure TPO, Irgacure TPO, Irganox 819, JRCure 1108, L-TPO, Lucirin 8893X, Lucirin LR 8728, Lucirin LR 8893, Lucirin LR 8953, Lucirin LR 8953X, Lucirin TPO, Lucirin TPO Solid, Lucirin TPO-X, Micure TPO, Omnirad TPO, Photocure TPO, Speedcure TPO, TPO, TPO 173, TPO-X, UV-Cure APO |
| InChI Formula | InChI=1S/C22H21O2P/c1-16-14-17(2)21(18(3)15-16)22(23)25(24,19-10-6-4-7-11-19)20-12-8-5-9-13-20/h4-15H,1-3H3 |
| SMILES | CC1=CC(=C(C(=C1)C)C(=O)P(=O)(C2=CC=CC=C2)C3=CC=CC=C3)C |
| IUPAC Name | diphenylphosphoryl-(2,4,6-trimethylphenyl)methanone |
| Molecular Weight (g/mol) | 348.37 |
| Formula Weight | 348.1279 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?